ENA405194636 Enamine

(2,5-dichlorophenyl)methanethiol

  • MDL number MFCD21187730
  • Cas Number : 17799-02-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134638

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134638
Availability details for
Quantity
  • Availability
  • Add to Cart