ENA405194638 Enamine

methyl 2-amino-3-(propan-2-yl)benzoate

  • MDL number MFCD12159858
  • Cas Number : 1178649-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134640

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134640
Availability details for
Quantity
  • Availability
  • Add to Cart