ENA405194641 Enamine

ethyl 2-(1H-pyrrol-1-yl)-4H,5H,7H-thieno[2,3-c]thiopyran-3-carboxylate

  • MDL number MFCD25970460
  • Cas Number : 1333824-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94...96
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134644

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134644
Availability details for
Quantity
  • Availability
  • Add to Cart