ENA405194642 Enamine

2-(2-phenylethyl)oxirane

  • MDL number MFCD01657147
  • Cas Number : 1126-76-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134645

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134645
Availability details for
Quantity
  • Availability
  • Add to Cart