ENA405194644 Enamine

5-amino-1,2-dimethyl-1H-imidazole-4-carboxamide

  • MDL number MFCD02752921
  • Cas Number : 78750-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130371
Availability details for
Quantity
  • Availability
  • Add to Cart