ENA405194647 Enamine

4-Amino-5-trifluoromethyl-4H-[1,2,4]triazole-3-thiol

  • MDL number MFCD00101076
  • Cas Number : 24848-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-03103

Safety Information

msds https://www.enaminestore.com/catalog/EN300-03103
Availability details for
Quantity
  • Availability
  • Add to Cart