ENA405194653 Enamine

4-(1H-imidazol-1-yl)thiophene-2-carbaldehyde

  • MDL number MFCD08668346
  • Cas Number : 91163-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123...125
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130429

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130429
Availability details for
Quantity
  • Availability
  • Add to Cart