ENA405194659 Enamine

3-bromo-5-phenyl-1,2,4-oxadiazole

  • MDL number MFCD06795878
  • Cas Number : 23432-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50...52
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134702

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134702
Availability details for
Quantity
  • Availability
  • Add to Cart