ENA405194665 Enamine

3-(hydroxymethyl)-1-methyl-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one

  • MDL number MFCD11109908
  • Cas Number : 1260656-84-5
  • Molecular Weight : 180.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(hydroxymethyl)-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-ol
Inchi Code 1S/C7H8N4O2/c1-11-6-5(4(2-12)10-11)7(13)9-3-8-6/h3,12H,2H2,1H3,(H,8,9,13)
Inchi Key WVBOGVTUZWACBM-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-134709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134709
Availability details for
Quantity
  • Availability
  • Add to Cart