ENA405194667 Enamine

O-[(4-fluorophenyl)methyl]hydroxylamine

  • MDL number MFCD11594367
  • Cas Number : 1782-40-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134714

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134714
Availability details for
Quantity
  • Availability
  • Add to Cart