ENA405194673 Enamine

1-benzyl-2-ethylpiperazine

  • MDL number MFCD07367738
  • Cas Number : 1031926-99-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134761

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134761
Availability details for
Quantity
  • Availability
  • Add to Cart