ENA405194674 Enamine

4-(2-aminobutyl)phenol hydrochloride

  • MDL number MFCD00051432
  • Cas Number : 78108-17-5
  • Molecular Weight : 201.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134773

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134773
Availability details for
Quantity
  • Availability
  • Add to Cart