ENA405194676 Enamine

2-(1H-imidazol-1-yl)pyridine-3-carboxylic acid

  • MDL number MFCD08668592
  • Cas Number : 954580-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130539

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130539
Availability details for
Quantity
  • Availability
  • Add to Cart