ENA405194679 Enamine

N-methyl-6-(morpholin-4-yl)pyridin-3-amine

  • MDL number MFCD17291955
  • Cas Number : 1342739-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 87...89
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134791

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134791
Availability details for
Quantity
  • Availability
  • Add to Cart