ENA405194683 Enamine

2-{[(2-methylbutan-2-yl)oxy]methyl}aniline

  • MDL number MFCD11196370
  • Cas Number : 1039930-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134796

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134796
Availability details for
Quantity
  • Availability
  • Add to Cart