ENA405194686 Enamine

N-{2-[(propan-2-yl)amino]ethyl}acetamide

  • MDL number MFCD11162574
  • Cas Number : 4814-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134803
Availability details for
Quantity
  • Availability
  • Add to Cart