ENA405194695 Enamine

1-(3,4-difluorophenyl)-2,2-dimethoxyethan-1-ol

  • MDL number MFCD21661933
  • Cas Number : 1461708-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134829

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134829
Availability details for
Quantity
  • Availability
  • Add to Cart