ENA405194696 Enamine

5-ethyl-1,3-thiazole-2-carbaldehyde

  • MDL number MFCD17251113
  • Cas Number : 339989-68-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130547
Availability details for
Quantity
  • Availability
  • Add to Cart