ENA405194698 Enamine

4-amino-1-(3-fluorophenyl)cyclohexan-1-ol

  • MDL number MFCD21321355
  • Cas Number : 1461708-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107...109
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134833

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134833
Availability details for
Quantity
  • Availability
  • Add to Cart