ENA405194709 Enamine

1-(3-bromopropyl)-4-methylbenzene

  • MDL number MFCD09744344
  • Cas Number : 54540-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130636

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130636
Availability details for
Quantity
  • Availability
  • Add to Cart