ENA405194713 Enamine

4-amino-1-methyl-4,5-dihydro-1H-1,2,4-triazol-5-one

  • MDL number MFCD25970480
  • Cas Number : 4254-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 152-154
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134867
Availability details for
Quantity
  • Availability
  • Add to Cart