ENA405194714 Enamine

1-(4-bromophenyl)-3,3,3-trifluoropropan-1-amine

  • MDL number MFCD09812854
  • Cas Number : 1016682-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134868
Availability details for
Quantity
  • Availability
  • Add to Cart