ENA405194717 Enamine

7-chloro-3-(oxan-4-yl)-[1,2,4]triazolo[4,3-c]pyrimidine

  • MDL number MFCD17243911
  • Cas Number : 1343379-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134871
Availability details for
Quantity
  • Availability
  • Add to Cart