ENA405194721 Enamine

3-phenyl-2-(phenylamino)propanoic acid

  • MDL number MFCD12899037
  • Cas Number : 23052-05-3
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134884

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134884
Availability details for
Quantity
  • Availability
  • Add to Cart