ENA405194723 Enamine

3-(5-chlorothiophen-2-yl)-3-oxopropanenitrile

  • MDL number MFCD07785835
  • Cas Number : 71683-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130760

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130760
Availability details for
Quantity
  • Availability
  • Add to Cart