ENA405194727 Enamine

2-(5-chloro-1H-1,2,4-triazol-3-yl)pyridine

  • MDL number MFCD19685601
  • Cas Number : 1315347-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 216...218
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134891

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134891
Availability details for
Quantity
  • Availability
  • Add to Cart