ENA405194730 Enamine

2-(1H-imidazol-1-yl)-5-nitropyridine

  • MDL number MFCD10699022
  • Cas Number : 23671-36-5
  • Molecular Weight : 190.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-134895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134895
Availability details for
Quantity
  • Availability
  • Add to Cart