ENA405194734 Enamine

6-methoxy-1,4-dihydroquinazolin-4-one

  • MDL number MFCD05662727
  • Cas Number : 19181-64-7
  • Molecular Weight : 176.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-130838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130838
Availability details for
Quantity
  • Availability
  • Add to Cart