ENA405194736 Enamine

2-amino-1-[3-(trifluoromethyl)phenyl]ethan-1-one hydrochloride

  • MDL number MFCD11974307
  • Cas Number : 61062-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 225-227
Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134924
Availability details for
Quantity
  • Availability
  • Add to Cart