ENA405194738 Enamine

methyl 2-chloro-2-(oxolan-3-yl)acetate

  • MDL number MFCD16694226
  • Cas Number : 1248819-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134926
Availability details for
Quantity
  • Availability
  • Add to Cart