ENA405194740 Enamine

methyl 2-(cyclopentylamino)benzoate

  • MDL number MFCD11179003
  • Cas Number : 1094286-24-4
  • Molecular Weight : 219.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134953
Availability details for
Quantity
  • Availability
  • Add to Cart