ENA405194741 Enamine

3-chloropyridine-4-carbohydrazide

  • MDL number MFCD17215778
  • Cas Number : 855636-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150...152
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134966

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134966
Availability details for
Quantity
  • Availability
  • Add to Cart