ENA405194754 Enamine

2-bromo-5-[(4-fluorophenyl)methyl]-1,3,4-thiadiazole

  • MDL number MFCD18345087
  • Cas Number : 1343894-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-134987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-134987
Availability details for
Quantity
  • Availability
  • Add to Cart