ENA405194756 Enamine

1-hydroxy-6-oxo-1,6-dihydropyridine-3-carboxylic acid

  • MDL number MFCD16485313
  • Cas Number : 677763-18-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 245...247
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134989

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134989
Availability details for
Quantity
  • Availability
  • Add to Cart