ENA405194759 Enamine

1-(4-amino-2,3-dihydro-1H-indol-1-yl)-2-methoxyethan-1-one

  • MDL number MFCD20318917
  • Cas Number : 1339774-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134992

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134992
Availability details for
Quantity
  • Availability
  • Add to Cart