ENA405194760 Enamine

1-methanesulfonyl-2,3-dihydro-1H-indol-4-amine

  • MDL number MFCD20302687
  • Cas Number : 1339796-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105...107
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-134993

Safety Information

msds https://www.enaminestore.com/catalog/EN300-134993
Availability details for
Quantity
  • Availability
  • Add to Cart