ENA405194769 Enamine

3-fluoro-4-(2,2,2-trifluoroethoxy)benzaldehyde

  • MDL number MFCD11156439
  • Cas Number : 502486-73-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135008

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135008
Availability details for
Quantity
  • Availability
  • Add to Cart