ENA405194777 Enamine

methyl 2-(3-bromophenyl)-2-oxoacetate

  • MDL number MFCD11973930
  • Cas Number : 81316-36-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 53...55
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130964

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130964
Availability details for
Quantity
  • Availability
  • Add to Cart