ENA405194778 Enamine

methyl 2-(3-bromophenyl)acetate

  • MDL number MFCD06858764
  • Cas Number : 150529-73-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130965

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130965
Availability details for
Quantity
  • Availability
  • Add to Cart