ENA405194781 Enamine

1-hydroxy-5,6,7,8-tetrahydronaphthalene-2-carboxylic acid

  • MDL number MFCD20638931
  • Cas Number : 103986-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135056

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135056
Availability details for
Quantity
  • Availability
  • Add to Cart