ENA405194786 Enamine

3-(morpholin-4-yl)pyridin-2-amine

  • MDL number MFCD11848423
  • Cas Number : 1036645-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135061

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135061
Availability details for
Quantity
  • Availability
  • Add to Cart