ENA405194802 Enamine

4-(1,5-dimethyl-1H-pyrazol-4-yl)pyrrolidin-2-one

  • MDL number MFCD21959960
  • Cas Number : 1461714-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135122
Availability details for
Quantity
  • Availability
  • Add to Cart