ENA405194803 Enamine

2-(4-carbamoylphenyl)acetic acid

  • MDL number MFCD20639603
  • Cas Number : 2393-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 238...240
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135123

Safety Information

msds https://www.enaminestore.com/catalog/EN300-135123
Availability details for
Quantity
  • Availability
  • Add to Cart