ENA405194804 Enamine

(4-chloro-3-methylphenyl)methanamine

  • MDL number MFCD18072521
  • Cas Number : 1155357-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-135124

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135124
Availability details for
Quantity
  • Availability
  • Add to Cart