ENA405194807 Enamine

1-methyl-8-oxa-2-azaspiro[4.5]decane

  • MDL number MFCD16737918
  • Cas Number : 1250678-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135127

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135127
Availability details for
Quantity
  • Availability
  • Add to Cart