ENA405194808 Enamine

3-(4-chlorobenzoyl)-1-methyl-1H-pyrazole

  • MDL number MFCD21179708
  • Cas Number : 325829-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135128
Availability details for
Quantity
  • Availability
  • Add to Cart