ENA405194811 Enamine

3-cyclohexyl-2-methylpropan-1-ol

  • MDL number MFCD20385961
  • Cas Number : 76019-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-130996

Safety Information

msds https://www.enaminestore.com/catalog/EN300-130996
Availability details for
Quantity
  • Availability
  • Add to Cart