ENA405194814 Enamine

4-(4-chlorophenyl)-2-methyl-4-oxobutanoic acid

  • MDL number MFCD01863260
  • Cas Number : 52240-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135168

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135168
Availability details for
Quantity
  • Availability
  • Add to Cart