ENA405194818 Enamine

1-{5-[(morpholin-4-yl)methyl]furan-2-yl}methanamine

  • MDL number MFCD06803230
  • Cas Number : 633335-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-135175

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-135175
Availability details for
Quantity
  • Availability
  • Add to Cart