ENA405194820 Enamine

1-(3,4-dichlorophenyl)-1H-1,2,3-triazole-4-carbaldehyde

  • MDL number MFCD21715559
  • Cas Number : 1325724-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-130999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130999
Availability details for
Quantity
  • Availability
  • Add to Cart